Assay Details
Target Gene Details
Entrez Gene ID: | 116848 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2A |
Gene Aliases: |
AA415431, C030005G16Rik, C78388, Tip5, Walp3, mKIAA0314 |
Location: |
Chr.10:128091056-128129304 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baz2a | NM_054078.2 | 4 | 680 | NP_473419.2 |
| AJ309544.2 | 2 | 414 | CAC69992.1 | |
| AK035400.1 | 4 | 777 | ||
| AK133149.1 | 4 | 680 | BAE21532.1 | |
| AK140355.1 | 5 | 1204 | ||
| AK155523.1 | 3 | 695 | BAE33307.1 | |
| BC020083.1 | 4 | 754 | ||
| BC038012.1 | 3 | 693 | AAH38012.1 | |
| BC048415.1 | 4 | 680 | AAH48415.1 |
More Information
Set Membership: |
|

Genomic Map