Assay Details
Target Gene Details
Entrez Gene ID: | 68281 |
Gene Name: | RIKEN cDNA 4930430F08 gene |
Gene Aliases: |
4930571E09Rik, A730088G13Rik, AW551239, C430008C19Rik |
Location: |
Chr.10:100572274-100589390 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930430F08Rik | NM_175128.2 | 7 | 2181 | NP_780337.2 |
| AK032226.1 | 7 | 2137 | BAC27770.1 | |
| AK036647.1 | 7 | 2164 | BAE43296.1 | |
| AK160693.1 | 7 | 2164 | ||
| BC082587.1 | 7 | 2131 | AAH82587.1 |
Target Gene Details
Entrez Gene ID: | 216274 |
Gene Name: | centrosomal protein 290 |
Gene Aliases: |
BC004690, Nphp6, b2b1454Clo, b2b1752Clo |
Location: |
Chr.10:100487548-100573655 on Build GRCm38 |
Assay Gene Location: | Within Intron 52 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep290 | NM_146009.2 | NP_666121.2 | ||
| AK029960.1 | BAC26700.1 | |||
| AK172940.1 | BAD32218.1 | |||
| BC004690.1 |
More Information
Set Membership: |
|

Genomic Map