Assay Details
Target Gene Details
Entrez Gene ID: | 75317 |
Gene Name: | PARP1 binding protein |
Gene Aliases: |
4930547N16Rik, AI503644, PARI |
Location: |
Chr.10:88091398-88146945 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parpbp | NM_029249.2 | 1 | 228 | NP_083525.1 |
| AK131981.1 | 1 | 214 | BAE20920.1 | |
| AK137160.1 | 1 | 177 | ||
| AK150571.1 | 1 | 192 | BAE29666.1 | |
| AK151268.1 | 1 | 192 | BAE30255.1 | |
| AK152208.1 | 1 | 192 | BAE31036.1 | |
| BC070476.1 | 1 | 228 |

Genomic Map