Assay Details
Target Gene Details
Entrez Gene ID: | 103220 |
Gene Name: | tetratricopeptide repeat domain 41 |
Gene Aliases: |
AI449705, AU041509, Gnn |
Location: |
Chr.10:86705632-86776844 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc41 | NM_001003910.2 | 6 | 1962 | NP_001003910.2 |
| NM_153595.3 | 5 | 1755 | NP_705823.3 | |
| AK133195.1 | 6 | 1903 | ||
| AY651020.1 | 5 | 1602 | AAT81232.1 | |
| AY651021.1 | 5 | 1602 | AAT81233.1 |

Genomic Map