Assay Details
Target Gene Details
Entrez Gene ID: | 208666 |
Gene Name: | DIRAS family, GTP-binding RAS-like 1 |
Gene Aliases: |
Di-Ras1, Gbts1, RIG |
Location: |
Chr.10:81019589-81025821 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Diras1 | NM_145217.2 | 2 | 1484 | NP_660252.1 |
| AK081336.1 | 2 | 1483 | ||
| BC057556.1 | 2 | 1484 | AAH57556.1 |
More Information
Set Membership: |
|

Genomic Map