Assay Details
Target Gene Details
Entrez Gene ID: | 74330 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 14 |
Gene Aliases: |
5730551F12Rik, DNAJ, DRIP78, HDJ3, LIP6 |
Location: |
Chr.10:128805468-128819446 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc14 | NM_028873.4 | 7 | 2353 | NP_083149.3 |
| AK032681.1 | 7 | 2335 | ||
| AK045345.1 | 7 | 2410 | BAC32320.1 | |
| AK045445.1 | 7 | 2447 | BAC32372.1 | |
| AK082579.1 | 7 | 2354 | BAC38536.1 | |
| AK083878.1 | 7 | 2365 | ||
| AK159388.1 | 7 | 2366 | BAE35043.1 |
Target Gene Details
Entrez Gene ID: | 66844 |
Gene Name: | ORM1-like 2 (S. cerevisiae) |
Gene Aliases: |
0610012C09Rik |
Location: |
Chr.10:128817457-128821632 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ormdl2 | NM_024180.5 | 4 | 1942 | NP_077142.1 |
| AV584065.1 | 1 | 114 |
More Information
Set Membership: |
|

Genomic Map