Assay Details
Target Gene Details
Entrez Gene ID: | 319415 |
Gene Name: | heparan sulfate (glucosamine) 3-O-sulfotransferase 5 |
Gene Aliases: |
D930005L05Rik, Gm1151, Hs3ost5 |
Location: |
Chr.10:36506804-36834397 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hs3st5 | NM_001081208.2 | 4 | 1416 | NP_001074677.1 |
| NM_001253355.1 | 6 | 1580 | NP_001240284.1 | |
| NM_001253356.1 | 5 | 1521 | NP_001240285.1 | |
| AK031910.1 | 6 | 1570 | BAC27601.1 | |
| BC147490.1 | 6 | 1573 | ||
| BC147493.1 | 5 | 1514 | ||
| BC150973.1 | 6 | 1573 | ||
| BC158004.1 | 6 | 1573 |
More Information
Set Membership: |
|

Genomic Map