Assay Details
Target Gene Details
Entrez Gene ID: | 216157 |
Gene Name: | transmembrane protein 259 |
Gene Aliases: |
AC087114.7gm2, ORF61, R32184_3, mFLJ00277 |
Location: |
Chr.10:79977120-79984330 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem259 | NM_001003949.3 | 11 | 1619 | NP_001003949.2 |
| AY096036.1 | 11 | 1329 | AAM34492.1 | |
| AY096037.1 | 10 | 1401 | AAM34493.1 | |
| BC005494.1 | 10 | 1142 | AAH05494.1 | |
| BC052787.1 | 10 | 1161 | AAH52787.1 | |
| BC100297.1 | 8 | 756 | AAI00298.1 |
More Information
Set Membership: |
|

Genomic Map