Assay Details
Target Gene Details
Entrez Gene ID: | 66844 |
Gene Name: | ORM1-like 2 (S. cerevisiae) |
Gene Aliases: |
0610012C09Rik |
Location: |
Chr.10:128817457-128821632 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ormdl2 | NM_024180.5 | 2 | 178 | NP_077142.1 |
AK002583.1 | 2 | 141 | BAB22207.1 | |
AK002824.1 | BAC25010.1 | |||
AK007548.1 | BAB25101.1 | |||
AK012681.1 | BAB28407.1 | |||
AK013455.1 | 2 | 151 | BAB28864.1 | |
BC002146.1 | 2 | 199 | AAH02146.1 | |
DV065903.1 | 2 | 221 |
