Assay Details
Target Gene Details
Entrez Gene ID: | 19279 |
Gene Name: | protein tyrosine phosphatase, receptor type, R |
Gene Aliases: |
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213 |
Location: |
Chr.10:116018200-116274932 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprr | NM_001161837.1 | 13 | 2656 | NP_001155309.1 |
NM_001161838.1 | 10 | 2613 | NP_001155310.1 | |
NM_001161839.1 | 11 | 2304 | NP_001155311.1 | |
NM_001161840.1 | 11 | 2496 | NP_001155312.1 | |
NM_011217.2 | 14 | 3262 | NP_035347.1 | |
AF041866.1 | 11 | 2421 | AAD09171.1 | |
D31898.1 | 14 | 3225 | BAA06696.1 | |
Z30313.1 | 13 | 2651 | CAA82957.1 |
