Assay Details
Target Gene Details
Entrez Gene ID: | 212898 |
Gene Name: | dermatan sulfate epimerase |
Gene Aliases: |
6030499O08, AI480506, B130024B19Rik, DS-epi1, Sart2 |
Location: |
Chr.10:34151386-34208275 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dse | NM_172508.2 | 6 | 2068 | NP_766096.1 |
| AK031737.1 | 6 | 1755 | ||
| AK045065.1 | 6 | 1975 | BAC32205.1 | |
| AK150737.1 | 6 | 2232 | ||
| AK153326.1 | 6 | 1741 | BAE31905.1 | |
| AK196259.1 | 1 | 98 | ||
| AK196274.1 | 1 | 98 | ||
| AK202322.1 | 1 | 98 | ||
| AK203692.1 | 1 | 98 | ||
| AK204558.1 | 1 | 98 | ||
| BC053095.1 | 6 | 2068 | AAH53095.1 |

Genomic Map