Assay Details
Target Gene Details
Entrez Gene ID: | 100038566 |
Gene Name: | predicted gene 10658 |
Gene Aliases: |
ENSMUSG00000074284 |
Location: |
Chr.9:57056897-57071966 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10658 | NR_045886.1 | |||
| AK085949.1 | ||||
| AK143982.1 | BAE25644.1 | |||
| AK162906.1 | BAE37109.1 | |||
| BU696022.1 |
Target Gene Details
Entrez Gene ID: | 56294 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 9 |
Gene Aliases: |
MEG2 |
Location: |
Chr.9:56994968-57062808 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn9 | NM_019651.2 | 13 | 2809 | NP_062625.2 |
| AF013490.1 | 13 | 2279 | AAB66898.1 | |
| AK089069.1 | 13 | 2383 | ||
| AK219745.1 | 1 | 341 | ||
| BC053017.1 | 13 | 2809 | AAH53017.1 |
More Information
Set Membership: |
|

Genomic Map