Assay Details
Target Gene Details
Entrez Gene ID: | 76832 |
Gene Name: | hydrolethalus syndrome 1 |
Gene Aliases: |
3010015K02Rik |
Location: |
Chr.9:35560821-35570069 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hyls1 | NM_029762.1 | NP_084038.1 | ||
| AK013896.1 | BAB29044.1 | |||
| AK083607.1 | 2 | 3665 | ||
| AK131639.1 | BAE20733.1 | |||
| BC035953.1 | AAH35953.1 | |||
| BY109268.1 |
Target Gene Details
Entrez Gene ID: | 67049 |
Gene Name: | pseudouridine synthase 3 |
Gene Aliases: |
2610020J05Rik, 5730412F04Rik |
Location: |
Chr.9:35558597-35567400 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pus3 | NM_023292.4 | 2 | 332 | NP_075781.3 |
| AF266505.1 | 1 | 286 | AAF91402.1 | |
| AK030639.1 | 3 | 636 | ||
| AK079095.1 | 2 | 333 | BAC37536.1 | |
| AK213612.1 | ||||
| BC029253.1 | 1 | 293 | AAH29253.1 |
More Information
Set Membership: |
|

Genomic Map