Assay Details
Target Gene Details
Entrez Gene ID: | 268281 |
Gene Name: | SNF2 histone linker PHD RING helicase |
Gene Aliases: |
2610103K11Rik, AA450458, AU024614, BC006883, D230017O13Rik, E130018M05 |
Location: |
Chr.10:11149321-11217595 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shprh | NM_001077707.1 | 2 | 707 | NP_001071175.1 |
| NM_001284354.1 | 2 | 707 | NP_001271283.1 | |
| NM_172937.3 | 2 | 707 | NP_766525.3 | |
| AK011809.1 | 2 | 632 | ||
| AK048265.1 | 2 | 665 | ||
| AK053448.1 | 2 | 708 | BAC35389.1 | |
| AK082160.1 | 2 | 711 | BAC38428.1 | |
| AK085285.1 | 2 | 645 | ||
| AY162264.1 | 2 | 613 | AAO26654.1 | |
| AY162266.1 | 2 | 613 | AAO26656.1 | |
| BC055003.1 | 2 | 678 | AAH55003.1 |
More Information
Set Membership: |
|

Genomic Map