Assay Details
Target Gene Details
Entrez Gene ID: | 20503 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 7 |
Gene Aliases: |
4921534N07Rik, Mct2 |
Location: |
Chr.10:125219284-125389562 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc16a7 | NM_011391.1 | 7 | 1880 | NP_035521.1 |
| AF058054.1 | 7 | 1880 | AAC13719.1 | |
| AK076612.1 | 6 | 1702 | BAC36415.1 | |
| AK085398.1 | 6 | 2232 | ||
| BC117803.1 | 6 | 1644 | ||
| BC117804.1 | 6 | 1644 | ||
| BC126960.1 | 6 | 1644 | ||
| CT010191.1 | CAJ18399.1 |
More Information
Set Membership: |
|

Genomic Map