Assay Details
Target Gene Details
Entrez Gene ID: | 100040851 |
Gene Name: | predicted gene 9767 |
Gene Aliases: |
ENSMUSG00000039098 |
Location: |
Chr.10:26078255-26079447 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm9767 | NR_028030.2 | 1 | 637 | |
| AK021041.1 | 1 | 638 | BAB32285.1 |
Target Gene Details
Entrez Gene ID: | 77220 |
Gene Name: | transmembrane protein 200A |
Gene Aliases: |
C030003D03Rik |
Location: |
Chr.10:25991182-26079408 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem200a | NM_029881.3 | 1 | 162 | NP_084157.2 |
| AK048673.1 | 1 | 73 | BAC33418.1 | |
| AK173299.1 | 1 | 162 | BAD32577.1 |

Genomic Map