Assay Details
Target Gene Details
Entrez Gene ID: | 77220 |
Gene Name: | transmembrane protein 200A |
Gene Aliases: |
C030003D03Rik |
Location: |
Chr.10:25991182-26079408 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem200a | NM_029881.3 | 2 | 1449 | NP_084157.2 |
| AK032569.1 | 2 | 1049 | BAC27929.1 | |
| AK048673.1 | 3 | 844 | BAC33418.1 | |
| AK165025.1 | 2 | 616 | ||
| AK173299.1 | 2 | 1449 | BAD32577.1 | |
| BC141266.1 | 2 | 674 | ||
| BC141267.1 | 2 | 674 |

Genomic Map