Assay Details
Target Gene Details
Entrez Gene ID: | 192653 |
Gene Name: | tetratricopeptide repeat domain 36 |
Gene Aliases: |
- |
Location: |
Chr.9:44799400-44802951 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc36 | NM_138951.1 | 3 | 712 | NP_620401.1 |
| AK149384.1 | 3 | 743 | BAE28845.1 | |
| AK165811.1 | 3 | 610 | BAE38390.1 | |
| BC021608.1 | 3 | 624 | AAH21608.1 | |
| BC021613.1 | 3 | 718 | AAH21613.1 | |
| BC024432.1 | 3 | 712 | AAH24432.1 | |
| BC026825.1 | 3 | 659 | AAH26825.1 |

Genomic Map