Assay Details
Target Gene Details
Entrez Gene ID: | 11540 |
Gene Name: | adenosine A2a receptor |
Gene Aliases: |
A2AAR, A2aR, AA2AR |
Location: |
Chr.10:75316860-75334792 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adora2a | NM_009630.3 | 3 | 1127 | NP_033760.2 |
| AK037787.1 | 2 | 349 | BAC29871.1 | |
| BC110692.1 | 2 | 627 | AAI10693.1 | |
| U05672.1 | 2 | 851 | AAA19000.1 | |
| Y13346.1 | 2 | 612 | CAA73788.1 |
More Information
Set Membership: |
|

Genomic Map