Assay Details
Target Gene Details
Entrez Gene ID: | 71902 |
Gene Name: | cullin associated and neddylation disassociated 1 |
Gene Aliases: |
2310038O07Rik, 6330512O03Rik, AI195005, AI846556, D10Ertd516e, Tp120a, mKIAA0829 |
Location: |
Chr.10:119198812-119240055 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cand1 | NM_027994.1 | 1 | 134 | NP_082270.1 |
| AK129225.1 | 1 | 95 | BAC98035.1 | |
| AK144869.1 | 1 | 126 | BAE26109.1 | |
| AK158080.1 | 1 | 130 |
More Information
Set Membership: |
|

Genomic Map