Assay Details
Target Gene Details
Entrez Gene ID: | 58802 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 4 |
Gene Aliases: |
1700058G18Rik, 2900045G12Rik |
Location: |
Chr.10:116417868-116473882 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnmb4 | NM_021452.1 | 3 | 737 | NP_067427.1 |
| AF215892.1 | 3 | 737 | AAF75597.1 | |
| AK013647.1 | 3 | 1096 | ||
| BC117779.2 |
Target Gene Details
Entrez Gene ID: | 67342 |
Gene Name: | potassium large conductance calcium-activated channel, subfamily M, beta member 4, opposite strand 1 |
Gene Aliases: |
1700058G18Rik |
Location: |
Chr.10:116418121-116421235 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnmb4os1 | NR_028107.1 | |||
| AK006831.1 | BAB24762.1 |
More Information
Set Membership: |
|

Genomic Map