Assay Details
Target Gene Details
Entrez Gene ID: | 338365 |
Gene Name: | solute carrier family 41, member 2 |
Gene Aliases: |
A230035L05Rik, SLC41A1-L1 |
Location: |
Chr.10:83230848-83338329 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc41a2 | NM_177388.3 | 11 | 3087 | NP_796362.1 |
| AK033822.1 | 1 | 1280 | ||
| AK038549.1 | 11 | 3088 | BAC30038.1 |
More Information
Set Membership: |
|

Genomic Map