Assay Details
Target Gene Details
Entrez Gene ID: | 22110 |
Gene Name: | testis-specific protein, Y-encoded-like 1 |
Gene Aliases: |
Tspyl |
Location: |
Chr.10:34282190-34284885 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspyl1 | NM_009433.3 | 1 | 1797 | NP_033459.1 |
| AK053812.1 | 1 | 1748 | BAC35536.1 | |
| AK159432.1 | 1 | 1778 | BAE35079.1 | |
| AK166438.1 | 1 | 1776 | ||
| AK166805.1 | 1 | 1726 | BAE39033.1 | |
| AK166893.1 | 1 | 1726 | BAE39098.1 | |
| BC011213.1 | 1 | 1744 | AAH11213.1 |
More Information
Set Membership: |
|

Genomic Map