Assay Details
Target Gene Details
Entrez Gene ID: | 237320 |
Gene Name: | aldehyde dehydrogenase 8 family, member A1 |
Gene Aliases: |
Raldh4 |
Location: |
Chr.10:21377300-21396578 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh8a1 | NM_178713.4 | 5 | 838 | NP_848828.1 |
| AF510322.1 | 5 | 847 | AAO64246.1 | |
| AK050298.1 | 5 | 839 | BAC34173.1 | |
| AK143752.1 | 5 | 839 | BAE25524.1 | |
| BC013511.1 | 3 | 380 | AAH13511.1 | |
| BC038493.1 | 5 | 850 | AAH38493.1 |
More Information
Set Membership: |
|

Genomic Map