Assay Details
Target Gene Details
Entrez Gene ID: | 52713 |
Gene Name: | coiled-coil domain containing 59 |
Gene Aliases: |
2300004H16Rik, D10Ertd718e |
Location: |
Chr.10:105841479-105847510 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc59 | NM_025602.3 | 2 | 380 | NP_079878.1 |
| AK009028.1 | 2 | 747 | BAB26036.1 | |
| AK012917.1 | 2 | 363 | BAB28548.1 | |
| AK019970.1 | 2 | 381 | BAB31941.1 | |
| AK047009.1 | 2 | 785 | ||
| AK135550.1 | 2 | 405 | BAE22578.1 | |
| BC027401.1 | 2 | 365 | AAH27401.1 |

Genomic Map