Assay Details
Target Gene Details
Entrez Gene ID: | 237339 |
Gene Name: | l(3)mbt-like 3 (Drosophila) |
Gene Aliases: |
AI481284, MBT-1 |
Location: |
Chr.10:26274470-26375989 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L3mbtl3 | NM_172787.2 | 22 | 2870 | NP_766375.1 |
| AK030412.1 | 9 | 1468 | BAC26951.1 | |
| AK045667.1 | 22 | 2871 | BAC32449.1 | |
| AK129443.1 | 18 | 2415 | BAC98253.1 | |
| BC082309.1 | 21 | 2740 | AAH82309.1 | |
| BC085192.1 | 21 | 2975 |
More Information
Set Membership: |
|

Genomic Map