Assay Details
Target Gene Details
Entrez Gene ID: | 50493 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
TR, TR1, TrxR1 |
Location: |
Chr.10:82833951-82897724 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Txnrd1 | NM_001042513.1 | 15 | 2831 | NP_001035978.1 |
| NM_001042514.1 | 14 | 2919 | NP_001035979.1 | |
| NM_001042523.1 | 15 | 3048 | NP_001035988.1 | |
| NM_015762.2 | 14 | 2724 | NP_056577.2 | |
| AB027565.1 | 14 | 2751 | BAA86985.2 | |
| AF333036.1 | 15 | 3048 | AAK01140.1 | |
| AK007891.1 | 1 | 164 | ||
| AK146125.1 | 14 | 2725 | BAE26918.1 | |
| AK151927.1 | ||||
| AK194198.1 | ||||
| AK199538.1 | ||||
| AK200909.1 | ||||
| AK202828.1 | ||||
| AK203123.1 | ||||
| AK204569.1 | 1 | 151 | ||
| AK210211.1 | ||||
| AK211837.1 | ||||
| AK217830.1 | ||||
| BC037643.1 | 14 | 3004 | AAH37643.1 |
More Information
Set Membership: |
|

Genomic Map