Assay Details
Target Gene Details
Entrez Gene ID: | 74443 |
Gene Name: | prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum) |
Gene Aliases: |
4933406E20Rik, AI853847, BB128974, Ph-4 |
Location: |
Chr.9:108578826-108597876 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P4htm | NM_028944.3 | 9 | 1572 | NP_083220.3 |
| AK016685.1 | 9 | 1546 | BAB30379.2 | |
| AK046783.1 | 9 | 1584 | BAC32866.1 | |
| AK047714.1 | 9 | 1572 | BAC33135.1 | |
| BC111871.1 | 9 | 1572 | AAI11872.1 |
More Information
Set Membership: |
|

Genomic Map