Assay Details
Target Gene Details
Entrez Gene ID: | 12569 |
Gene Name: | cyclin-dependent kinase 5, regulatory subunit 1 (p35) |
Gene Aliases: |
Cdk5r, D11Bwg0379e, p25, p35 |
Location: |
Chr.11:80477046-80481179 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5r1 | NM_009871.2 | 1 | 2063 | NP_034001.1 |
| AK039999.1 | 2 | 1854 | BAE20562.1 | |
| AK043203.1 | 1 | 2039 | BAC31490.1 | |
| AK048617.1 | 1 | 2042 | BAC33394.1 | |
| AK079557.1 | 1 | 2066 | BAC37683.1 | |
| AK084003.1 | 1 | 2019 | ||
| AK086621.1 | 1 | 2085 | BAC39703.1 | |
| AK141549.1 | 1 | 2076 | BAE24731.1 | |
| AK142868.1 | 1 | 2087 | ||
| AK169667.1 | 1 | 2067 | BAE41289.1 | |
| BC046823.1 | 1 | 2049 | AAH46823.1 | |
| BC050768.1 | 1 | 2063 | AAH50768.1 | |
| BC058697.1 | 1 | 2055 | AAH58697.1 |
More Information
Set Membership: |
|

Genomic Map