Assay Details
Target Gene Details
Entrez Gene ID: | 14457 |
Gene Name: | growth arrest specific 7 |
Gene Aliases: |
AW124766, B230343A10Rik, Gas7-cb |
Location: |
Chr.11:67455024-67688992 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas7 | NM_001109657.3 | 1 | 270 | NP_001103127.1 |
| NM_008088.5 | NP_032114.2 | |||
| AK017394.1 | 1 | 70 | ||
| AK031455.1 | 1 | 271 | ||
| AK076926.1 | ||||
| AK154463.1 | 1 | 73 | BAE32603.1 | |
| BB620887.1 | 1 | 271 | ||
| BC120528.1 | ||||
| BC120530.1 | ||||
| U19860.1 | AAA85259.1 |
More Information
Set Membership: |
|

Genomic Map