Assay Details
Target Gene Details
Entrez Gene ID: | 14799 |
Gene Name: | glutamate receptor, ionotropic, AMPA1 (alpha 1) |
Gene Aliases: |
2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1 |
Location: |
Chr.11:57011571-57330244 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gria1 | NM_001113325.2 | 1 | 351 | NP_001106796.1 |
| NM_008165.4 | 1 | 351 | NP_032191.2 | |
| AB102773.1 | ||||
| AB102774.1 | ||||
| AK049958.1 | 1 | 160 | ||
| BC056397.1 | AAH56397.1 | |||
| BC060702.1 | AAH60702.1 | |||
| CJ155313.1 | 1 | 279 | ||
| X57497.1 | 1 | 198 | CAA40734.1 |
More Information
Set Membership: |
|

Genomic Map