Assay Details
Target Gene Details
Entrez Gene ID: | 14799 |
Gene Name: | glutamate receptor, ionotropic, AMPA1 (alpha 1) |
Gene Aliases: |
2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1 |
Location: |
Chr.11:57011571-57330244 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gria1 | NM_001113325.2 | 11 | 2110 | NP_001106796.1 |
| NM_001252403.1 | 11 | 1737 | NP_001239332.1 | |
| NM_008165.4 | 11 | 2110 | NP_032191.2 | |
| AB102773.1 | 11 | 1694 | ||
| AB102774.1 | 11 | 1694 | ||
| AB104707.1 | 1 | 82 | ||
| AK049958.1 | 11 | 1920 | ||
| AK135107.1 | 11 | 1738 | ||
| BC056397.1 | 11 | 1772 | AAH56397.1 | |
| BC060702.1 | 11 | 1693 | AAH60702.1 | |
| X57497.1 | 11 | 1957 | CAA40734.1 |
More Information
Set Membership: |
|

Genomic Map