Assay Details
Target Gene Details
Entrez Gene ID: | 20848 |
Gene Name: | signal transducer and activator of transcription 3 |
Gene Aliases: |
1110034C02Rik, AW109958, Aprf |
Location: |
Chr.11:100886810-100939570 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stat3 | NM_011486.4 | 24 | 3784 | NP_035616.1 |
| NM_213659.2 | 24 | 3834 | NP_998824.1 | |
| NM_213660.2 | 24 | 3831 | NP_998825.1 | |
| AK004083.1 | 1 | 356 | ||
| AK153170.1 | 1 | 1014 | ||
| AK153472.1 | 24 | 3746 | BAE32023.1 |
More Information
Set Membership: |
|

Genomic Map