Assay Details
Target Gene Details
Entrez Gene ID: | 237911 |
Gene Name: | BRCA1 interacting protein C-terminal helicase 1 |
Gene Aliases: |
3110009N10Rik, 8030460J03Rik, Bach1, FACJ, Fancj, OF |
Location: |
Chr.11:86058131-86201267 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brip1 | NM_178309.2 | 19 | 2915 | NP_840094.1 |
| AK033205.1 | 19 | 2886 | ||
| AK041841.1 | 4 | 488 | BAE20606.1 | |
| AK080771.1 | 4 | 542 | BAC38016.1 | |
| BC094252.1 | 19 | 2864 | AAH94252.1 |
More Information
Set Membership: |
|

Genomic Map