Assay Details
Target Gene Details
Entrez Gene ID: | 216848 |
Gene Name: | chromodomain helicase DNA binding protein 3 |
Gene Aliases: |
2600010P09Rik, AF020312, Chd7, Prp7, Prp9-1 |
Location: |
Chr.11:69343273-69369426 on Build GRCm38 |
Assay Gene Location: | Within Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chd3 | NM_146019.4 | 40 | 6477 | NP_666131.3 |
| AK011183.1 | 3 | 678 | ||
| BC030435.1 | 7 | 1360 | AAH30435.1 | |
| BC055484.1 | 3 | 967 | AAH55484.1 |

Genomic Map