Assay Details
Target Gene Details
Entrez Gene ID: | 66569 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 1 |
Gene Aliases: |
2610020H15Rik |
Location: |
Chr.11:87033794-87074953 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdpd1 | NM_025638.2 | 10 | 2232 | NP_079914.1 |
| AK039183.1 | 1 | 1214 | ||
| AK167812.1 | 10 | 2215 | BAE39839.1 | |
| AK202723.1 | ||||
| BC016541.1 | 10 | 2155 | AAH16541.1 | |
| BY697784.1 | 1 | 146 | ||
| CA451251.1 | 1 | 537 |
More Information
Set Membership: |
|

Genomic Map