Assay Details
Target Gene Details
Entrez Gene ID: | 77605 |
Gene Name: | H2A histone family, member V |
Gene Aliases: |
C530002L11Rik, H2a.z-2, H2av |
Location: |
Chr.11:6427226-6444443 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2afv | NM_029938.1 | 5 | 840 | NP_084214.1 |
More Information
Set Membership: |
|

Genomic Map