Assay Details
Target Gene Details
Entrez Gene ID: | 18023 |
Gene Name: | nuclear factor, erythroid derived 2,-like 1 |
Gene Aliases: |
AA408798, AW212678, LCR-F1, Lcrf1, NRF1, TCF-11, TCF11 |
Location: |
Chr.11:96817413-96829968 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfe2l1 | NM_001130450.1 | 2 | 780 | NP_001123922.1 |
| NM_008686.3 | 2 | 838 | NP_032712.2 | |
| AF071084.1 | 1 | 184 | AAC83235.1 | |
| AK131183.1 | 2 | 775 | BAD21433.1 | |
| AK152040.1 | 2 | 819 | BAE30899.1 | |
| AK153529.1 | 2 | 819 | BAE32068.1 | |
| AK154167.1 | 2 | 750 | BAE32418.1 | |
| BC043063.1 | 1 | 420 | AAH43063.1 | |
| BC047283.1 | 1 | 180 | AAH47283.1 | |
| CN527202.1 | ||||
| X78709.1 | 1 | 216 | CAA55362.1 |
More Information
Set Membership: |
|

Genomic Map