Assay Details
Target Gene Details
Entrez Gene ID: | 56218 |
Gene Name: | POZ (BTB) and AT hook containing zinc finger 1 |
Gene Aliases: |
8430401L15Rik, AI447519, Mazr, Patz, Zfp278 |
Location: |
Chr.11:3289131-3309083 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Patz1 | NM_001253690.1 | 1 | 711 | NP_001240619.1 |
| NM_001253691.1 | 1 | 711 | NP_001240620.1 | |
| NM_019574.3 | 1 | 711 | NP_062520.1 | |
| NR_045108.2 | ||||
| NR_045109.2 | ||||
| AB029397.1 | 1 | 711 | BAA90874.1 | |
| AK018372.1 | ||||
| AK166189.1 | 1 | 832 | BAE38619.1 | |
| AK185454.1 | 1 | 75 | ||
| BC043035.1 | 1 | 729 | AAH43035.1 | |
| BC055947.1 | AAH55947.1 |
More Information
Set Membership: |
|

Genomic Map