Assay Details
Target Gene Details
Entrez Gene ID: | 104457 |
Gene Name: | RIKEN cDNA 0610010K14 gene |
Gene Aliases: |
1110020A23Rik, AL033328, AU045833, Bap18 |
Location: |
Chr.11:70235204-70237914 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 0610010K14Rik | NM_001177601.1 | 6 | 759 | NP_001171072.1 |
| NM_001177603.1 | 5 | 654 | NP_001171074.1 | |
| NM_001177606.1 | 4 | 552 | NP_001171077.1 | |
| NM_001177607.1 | 4 | 579 | NP_001171078.1 | |
| NM_026757.2 | 5 | 653 | NP_081033.1 | |
| NM_145758.1 | 6 | 755 | NP_665701.1 | |
| AA189586.1 | 3 | 265 | ||
| AK003842.1 | 5 | 653 | BAB23031.1 | |
| AK169173.1 | 4 | 543 | BAE40952.1 | |
| AK170380.1 | 5 | 652 | BAE41758.1 | |
| AW557771.2 | 1 | 124 | ||
| BC055357.1 | 5 | 594 | AAH55357.1 | |
| CB192659.1 | 5 | 655 | ||
| CK791552.1 |
More Information
Set Membership: |
|

Genomic Map