Assay Details
Target Gene Details
Entrez Gene ID: | 260302 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 3 |
Gene Aliases: |
C230037M19Rik, mKIAA0154 |
Location: |
Chr.11:115584255-115603917 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gga3 | NM_001252067.1 | 16 | 2660 | NP_001238996.1 |
| NM_173048.3 | 17 | 2894 | NP_766636.2 | |
| AK031086.1 | 17 | 2866 | BAC27246.1 | |
| AK082327.1 | 16 | 2624 | ||
| AK089029.1 | 1 | 162 | ||
| AK122212.2 | 17 | 2863 | BAC65494.2 | |
| AK132183.1 | 17 | 2862 | BAE21019.1 | |
| AK147882.1 | 17 | 2862 |

Genomic Map