Assay Details
Target Gene Details
Entrez Gene ID: | 18053 |
Gene Name: | nerve growth factor receptor (TNFR superfamily, member 16) |
Gene Aliases: |
LNGFR, Tnfrsf16, p75, p75NGFR, p75NTR |
Location: |
Chr.11:95568818-95587698 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ngfr | NM_033217.3 | 3 | 598 | NP_150086.2 |
| AF105292.1 | 3 | 456 | AAD17943.1 | |
| AK037248.1 | 3 | 599 | BAC29775.1 | |
| BB630380.1 | 3 | 599 | ||
| BC038365.1 | 3 | 589 | AAH38365.1 | |
| KC855565.1 |
More Information
Set Membership: |
|

Genomic Map