Assay Details
Target Gene Details
Entrez Gene ID: | 18053 |
Gene Name: | nerve growth factor receptor (TNFR superfamily, member 16) |
Gene Aliases: |
LNGFR, Tnfrsf16, p75, p75NGFR, p75NTR |
Location: |
Chr.11:95568818-95587698 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ngfr | NM_033217.3 | 4 | 889 | NP_150086.2 |
| AF105292.1 | 4 | 747 | AAD17943.1 | |
| AK037248.1 | 4 | 890 | BAC29775.1 | |
| BC038365.1 | 4 | 880 | AAH38365.1 |
More Information
Set Membership: |
|

Genomic Map