Assay Details
Target Gene Details
Entrez Gene ID: | 18559 |
Gene Name: | phosphatidylcholine transfer protein |
Gene Aliases: |
PC-TP, StarD2 |
Location: |
Chr.11:89983417-90002914 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pctp | NM_008796.2 | 6 | 1078 | NP_032822.2 |
| AF114437.1 | 4 | 783 | AAF08346.1 | |
| AK151328.1 | 1 | 381 |

Genomic Map