Assay Details
Target Gene Details
Entrez Gene ID: | 93896 |
Gene Name: | glucagon-like peptide 2 receptor |
Gene Aliases: |
9530092J08Rik, GLP-2 |
Location: |
Chr.11:67663299-67771153 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glp2r |
Target Gene Details
Entrez Gene ID: | 19674 |
Gene Name: | recoverin |
Gene Aliases: |
CAR, S-modulin |
Location: |
Chr.11:67695326-67703355 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcvrn | NM_009038.2 | 3 | 827 | NP_033064.1 |
| BC110564.2 | AAI10565.1 | |||
| CK617615.1 | 1 | 242 | ||
| CK627353.1 | 1 | 267 | ||
| X66196.1 | 3 | 835 | CAA46958.1 |

Genomic Map