Assay Details
Target Gene Details
Entrez Gene ID: | 170472 |
Gene Name: | RecQ protein-like 5 |
Gene Aliases: |
RecQ5, Recq5b, Recql5b |
Location: |
Chr.11:115892595-115933531 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Recql5 | NM_130454.2 | 15 | 2604 | NP_569721.1 |
| AB059999.1 | 15 | 2118 | BAB79232.1 | |
| AK049269.1 | 10 | 1004 | BAC33647.1 | |
| AK050965.1 | 13 | 1333 | BAC34479.1 | |
| BC019613.1 | 2 | 221 | AAH19613.1 |
More Information
Set Membership: |
|

Genomic Map