Assay Details
Target Gene Details
Entrez Gene ID: | 67338 |
Gene Name: | ring finger and FYVE like domain containing protein |
Gene Aliases: |
1700051E09Rik, 4930516L10Rik, BG080975, Carp2 |
Location: |
Chr.11:82803819-82871239 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rffl | NM_001007465.3 | 3 | 736 | NP_001007466.1 |
| NM_001164569.1 | 4 | 880 | NP_001158041.1 | |
| NM_001164570.1 | 4 | 822 | NP_001158042.1 | |
| NM_001164571.1 | 3 | 739 | NP_001158043.1 | |
| NM_026097.3 | 3 | 847 | NP_080373.1 | |
| AF434814.1 | 2 | 525 | AAL30769.1 | |
| AK006757.1 | 3 | 836 | ||
| AK007189.1 | 3 | 835 | BAB24891.1 | |
| AK015806.1 | 3 | 848 | BAB29984.1 | |
| AK028095.1 | 3 | 836 | BAC25744.1 | |
| AK128983.1 | 4 | 823 | BAC87665.1 | |
| AK150213.1 | 3 | 669 | BAE29383.1 | |
| AK150541.1 | 4 | 842 | BAE29644.1 | |
| AK159442.1 | 3 | 695 | ||
| AK169393.1 | 3 | 680 | BAE41140.1 | |
| AK169988.1 | 3 | 737 | ||
| AK179164.1 | 1 | 98 | ||
| BC118517.1 | 3 | 622 |
More Information
Set Membership: |
|

Genomic Map