Assay Details
Target Gene Details
Entrez Gene ID: | 118454 |
Gene Name: | gap junction protein, gamma 2 |
Gene Aliases: |
B230382L12Rik, Cx47, Gja12 |
Location: |
Chr.11:59175564-59183213 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjc2 | NM_080454.4 | 2 | 1469 | NP_536702.3 |
| NM_175452.4 | 2 | 1487 | NP_780661.2 | |
| AK046421.1 | 2 | 1488 | BAC32722.1 | |
| AK046609.1 | 2 | 1470 | BAC32806.1 | |
| BC113759.1 | 2 | 973 | AAI13760.1 |
More Information
Set Membership: |
|

Genomic Map