Assay Details
Target Gene Details
Entrez Gene ID: | 353155 |
Gene Name: | gap junction protein, delta 3 |
Gene Aliases: |
Gja11, Gjc1, cx30.2 |
Location: |
Chr.11:98982180-98983016 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjd3 | NM_178596.2 | 1 | 144 | NP_848711.2 |
More Information
Set Membership: |
|

Genomic Map