Assay Details
Target Gene Details
Entrez Gene ID: | 13650 |
Gene Name: | rhomboid family 1 (Drosophila) |
Gene Aliases: |
C16ORF8, Dist, Dist1, Egfr-rs, mKIAA4242 |
Location: |
Chr.11:32209584-32222331 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhbdf1 | NM_001291818.1 | 8 | 1155 | NP_001278747.1 |
| NM_010117.2 | 8 | 1274 | NP_034247.2 | |
| AK144437.1 | 8 | 1143 | ||
| AK220566.1 | 5 | 2690 | BAD90543.1 | |
| BC023469.1 | 3 | 422 | AAH23469.1 | |
| BC027346.1 | 8 | 1219 | AAH27346.1 |
More Information
Set Membership: |
|

Genomic Map